COM448669184 Combi-Blocks, Inc.

1-Ethyl-4-(6-fluoro-5-methylpyridin-3-yl)piperidin-4-ol

  • MDL number MFCD28166404
  • Cas Number : 1820674-37-0
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1996

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1996
Availability details for
Quantity
  • Availability
  • Add to Cart