COM448669187 Combi-Blocks, Inc.

Ethyl 5-BOC-6-methyl-4-oxopyrazolo[1,5-a]pyrazine-2-carboxylate

  • MDL number MFCD28143964
  • Cas Number : 1820647-45-7
  • Molecular Weight : 321.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1999

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-1999
Availability details for
Quantity
  • Availability
  • Add to Cart