COM448669199 Combi-Blocks, Inc.

Quinoline-4-carboxamide

  • MDL number MFCD00672908
  • Cas Number : 4363-95-5
  • Molecular Weight : 172.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2044

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2044
Availability details for
Quantity
  • Availability
  • Add to Cart