COM448669202 Combi-Blocks, Inc.

(3-methoxypyridin-4-yl)methanol

  • MDL number MFCD16606979
  • Cas Number : 102074-60-2
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2049

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2049
Availability details for
Quantity
  • Availability
  • Add to Cart