COM448669209 Combi-Blocks, Inc.

N-Methylpyrimidin-4-amine

  • MDL number MFCD10697837
  • Cas Number : 22632-10-6
  • Molecular Weight : 109.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2070

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2070
Availability details for
Quantity
  • Availability
  • Add to Cart