COM448669210 Combi-Blocks, Inc.

6-Bromo-7-chloro-2-methylquinazolin-4-ol

  • MDL number MFCD28397299
  • Cas Number : 1698026-73-1
  • Molecular Weight : 273.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2076

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QY-2076
Availability details for
Quantity
  • Availability
  • Add to Cart