COM448669263 Combi-Blocks, Inc.

1-(Cyclopropylmethyl)pyrazole

  • MDL number MFCD18889145
  • Cas Number : 1344382-51-9
  • Molecular Weight : 122.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5066

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5066
Availability details for
Quantity
  • Availability
  • Add to Cart