COM448669330 Combi-Blocks, Inc.

3-Hydroxy-2-methoxynaphthalene

  • MDL number MFCD03093901
  • Cas Number : 39110-92-4
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5138

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5138
Availability details for
Quantity
  • Availability
  • Add to Cart