COM448669682 Combi-Blocks, Inc.

N,N-Dimethylpyrrole-1-sulfonamide

  • MDL number MFCD28127204
  • Cas Number : 1820619-15-5
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 70
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5662

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5662
Availability details for
Quantity
  • Availability
  • Add to Cart