COM448669747 Combi-Blocks, Inc.

(2-Hydroxyphenyl)thiourea

  • MDL number MFCD00041159
  • Cas Number : 1520-26-9
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5742

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5742
Availability details for
Quantity
  • Availability
  • Add to Cart