COM448669748 Combi-Blocks, Inc.

(5-Chloro-2-hydroxyphenyl)thiourea

  • MDL number MFCD00041160
  • Cas Number : 89793-06-6
  • Molecular Weight : 202.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5744

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5744
Availability details for
Quantity
  • Availability
  • Add to Cart