COM448669749 Combi-Blocks, Inc.

(2-Hydroxy-5-methylphenyl)thiourea

  • MDL number MFCD13187595
  • Cas Number : 500284-61-7
  • Molecular Weight : 182.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5745

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5745
Availability details for
Quantity
  • Availability
  • Add to Cart