COM448669751 Combi-Blocks, Inc.

3-(4-Bromo-2-fluorophenyl)-5-(chloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD27981338
  • Cas Number : 1791431-60-1
  • Molecular Weight : 291.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5751

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5751
Availability details for
Quantity
  • Availability
  • Add to Cart