COM448669765 Combi-Blocks, Inc.

2-Chloro-5-fluoro-1,3-benzoxazole

  • MDL number MFCD11890748
  • Cas Number : 135533-78-7
  • Molecular Weight : 171.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5765

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5765
Availability details for
Quantity
  • Availability
  • Add to Cart