COM448669773 Combi-Blocks, Inc.

7-Chloro-4-methyl-2H-1,4-benzoxazin-3-one

  • MDL number MFCD26663865
  • Cas Number : 1508393-11-0
  • Molecular Weight : 197.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5774

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5774
Availability details for
Quantity
  • Availability
  • Add to Cart