COM448669775 Combi-Blocks, Inc.

[3-(2-Bromophenyl)-1,2,4-oxadiazol-5-yl]methanamine

  • MDL number MFCD09034254
  • Cas Number : 937665-72-0
  • Molecular Weight : 254.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5777

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5777
Availability details for
Quantity
  • Availability
  • Add to Cart