COM448669777 Combi-Blocks, Inc.

[3-(2-Bromobenzyl)-1,2,4-oxadiazol-5-yl]methanamine

  • MDL number MFCD17225871
  • Cas Number : 1341130-08-2
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5779

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5779
Availability details for
Quantity
  • Availability
  • Add to Cart