COM448669778 Combi-Blocks, Inc.

2-(5-Bromo-2-fluorophenyl)propan-2-ol

  • MDL number MFCD23759088
  • Cas Number : 1206774-43-7
  • Molecular Weight : 233.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5780

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5780
Availability details for
Quantity
  • Availability
  • Add to Cart