COM448669782 Combi-Blocks, Inc.

[3-(4-Bromobenzyl)-1,2,4-oxadiazol-5-yl]methanamine

  • MDL number MFCD11205871
  • Cas Number : 1094351-79-7
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5785

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5785
Availability details for
Quantity
  • Availability
  • Add to Cart