COM448669784 Combi-Blocks, Inc.

N-BOC-3-[(3-Bromobenzene)sulfonyl]azetidine

  • MDL number MFCD06657047
  • Cas Number : 887593-56-8
  • Molecular Weight : 376.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5788

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5788
Availability details for
Quantity
  • Availability
  • Add to Cart