COM448669791 Combi-Blocks, Inc.

3-[(2-Bromophenyl)methyl]-5-(chloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD17225868
  • Cas Number : 1339141-04-6
  • Molecular Weight : 287.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5798

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5798
Availability details for
Quantity
  • Availability
  • Add to Cart