COM448669794 Combi-Blocks, Inc.

2-(N-BOC-Azetidin-3-yl)sulfanylphenylboronic acid, pinacol ester

  • MDL number MFCD27981339
  • Cas Number : 2377610-91-6
  • Molecular Weight : 391.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5801

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5801
Availability details for
Quantity
  • Availability
  • Add to Cart