COM448669798 Combi-Blocks, Inc.

4-Fluoro-3-piperidinomethylphenylboronic acid pinacol ester

  • MDL number MFCD27981341
  • Cas Number : 2377611-14-6
  • Molecular Weight : 319.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5805

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5805
Availability details for
Quantity
  • Availability
  • Add to Cart