COM448669803 Combi-Blocks, Inc.

Ethyl 5-cyano-1-benzothiophene-2-carboxylate

  • MDL number MFCD13180921
  • Cas Number : 105191-14-8
  • Molecular Weight : 231.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5810

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5810
Availability details for
Quantity
  • Availability
  • Add to Cart