COM448669804 Combi-Blocks, Inc.

[3-(2-Fluorophenyl)-1,2,4-oxadiazol-5-yl]methanamine

  • MDL number MFCD08899396
  • Cas Number : 919017-55-3
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5811

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5811
Availability details for
Quantity
  • Availability
  • Add to Cart