COM448669807 Combi-Blocks, Inc.

Methyl 1-methylcyclohexane-1-carboxylate

  • MDL number MFCD12025265
  • Cas Number : 825-04-7
  • Molecular Weight : 156.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5815

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5815
Availability details for
Quantity
  • Availability
  • Add to Cart