COM448669815 Combi-Blocks, Inc.

Ethyl (2E)-3-(4-bromo-2-nitrophenyl)prop-2-enoate

  • MDL number MFCD24335259
  • Cas Number : 1094851-38-3
  • Molecular Weight : 300.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5825

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5825
Availability details for
Quantity
  • Availability
  • Add to Cart