COM448669816 Combi-Blocks, Inc.

4-(Isopropylaminomethyl)-3-fluorophenylboronic acid, pinacol ester

  • MDL number MFCD27981345
  • Molecular Weight : 293.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5826

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5826
Availability details for
Quantity
  • Availability
  • Add to Cart