COM448669817 Combi-Blocks, Inc.

Ethyl 6-chloro-1-benzothiophene-2-carboxylate

  • MDL number MFCD25372107
  • Cas Number : 105191-53-5
  • Molecular Weight : 240.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5827

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5827
Availability details for
Quantity
  • Availability
  • Add to Cart