COM448669819 Combi-Blocks, Inc.

{3-[3-(Trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}methanamine

  • MDL number MFCD10696390
  • Cas Number : 944905-99-1
  • Molecular Weight : 243.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5829

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5829
Availability details for
Quantity
  • Availability
  • Add to Cart