COM448669820 Combi-Blocks, Inc.

5-Bromo-1-benzofuran-2-carboxamide

  • MDL number MFCD06357123
  • Cas Number : 35351-21-4
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5830

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5830
Availability details for
Quantity
  • Availability
  • Add to Cart