COM448669821 Combi-Blocks, Inc.

[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]methanamine

  • MDL number MFCD07087661
  • Cas Number : 890324-74-0
  • Molecular Weight : 189.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5832

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5832
Availability details for
Quantity
  • Availability
  • Add to Cart