COM448669831 Combi-Blocks, Inc.

(2E)-3-(4-Bromo-2-fluorophenyl)prop-2-enamide

  • MDL number MFCD09910885
  • Cas Number : 568595-13-1
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5872

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5872
Availability details for
Quantity
  • Availability
  • Add to Cart