COM448669845 Combi-Blocks, Inc.

2-Ethoxy-3-methylpyridine-5-carboxylic acid

  • MDL number MFCD21087314
  • Cas Number : 1355216-85-1
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5889

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5889
Availability details for
Quantity
  • Availability
  • Add to Cart