COM448669847 Combi-Blocks, Inc.

Methyl 2-(4-bromophenyl)-2-cyanoacetate

  • MDL number MFCD19329148
  • Cas Number : 1218950-96-9
  • Molecular Weight : 254.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5891

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5891
Availability details for
Quantity
  • Availability
  • Add to Cart