COM448669850 Combi-Blocks, Inc.

Ethyl 6-methyl-1-benzothiophene-2-carboxylate

  • MDL number MFCD28405095
  • Cas Number : 82787-72-2
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5894

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5894
Availability details for
Quantity
  • Availability
  • Add to Cart