COM448669853 Combi-Blocks, Inc.

1-Bromo-4-ethoxy-2-methoxybenzene

  • MDL number MFCD23162082
  • Cas Number : 1353776-78-9
  • Molecular Weight : 231.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5897

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5897
Availability details for
Quantity
  • Availability
  • Add to Cart