COM448669858 Combi-Blocks, Inc.

3-Methyl-5-phenyl-1,2,4-oxadiazole

  • MDL number MFCD00464109
  • Cas Number : 1199-00-4
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5904

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5904
Availability details for
Quantity
  • Availability
  • Add to Cart