COM448669865 Combi-Blocks, Inc.

N-BOC-3-(benzylsulfonyl)azetidine

  • MDL number MFCD28143968
  • Cas Number : 1820687-96-4
  • Molecular Weight : 311.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5914

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5914
Availability details for
Quantity
  • Availability
  • Add to Cart