COM448669869 Combi-Blocks, Inc.

5-(2-Fluorophenyl)-3-methyl-1,2,4-oxadiazole

  • MDL number MFCD28166409
  • Cas Number : 1820747-54-3
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5920

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5920
Availability details for
Quantity
  • Availability
  • Add to Cart