COM448669876 Combi-Blocks, Inc.

(2E)-3-(3-fluoropyridin-4-yl)prop-2-enoic acid

  • MDL number MFCD28166411
  • Cas Number : 1567668-00-1
  • Molecular Weight : 167.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5928

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5928
Availability details for
Quantity
  • Availability
  • Add to Cart