COM448669879 Combi-Blocks, Inc.

4-Bromo-2-(ethoxymethyl)thiophene

  • MDL number MFCD16095116
  • Cas Number : 1065184-23-7
  • Molecular Weight : 221.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5931

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5931
Availability details for
Quantity
  • Availability
  • Add to Cart