COM448669920 Combi-Blocks, Inc.

5-Chloro-1-methylindazole

  • MDL number MFCD11977519
  • Cas Number : 1209268-02-9
  • Molecular Weight : 166.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?SH-5998
Availability details for
Quantity
  • Availability
  • Add to Cart