COM448669922 Combi-Blocks, Inc.

3-(2-Chlorophenyl)-4H-1,2,4-triazole

  • MDL number MFCD07352278
  • Cas Number : 56015-92-0
  • Molecular Weight : 179.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6000

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SH-6000
Availability details for
Quantity
  • Availability
  • Add to Cart