COM448670205 Combi-Blocks, Inc.

6-Benzyl-5,6,7,8-tetrahydro-2,6-naphthyridin-1(2h)-one

  • MDL number MFCD06762738
  • Cas Number : 601514-62-9
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-0294

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-0294
Availability details for
Quantity
  • Availability
  • Add to Cart