COM448671428 Combi-Blocks, Inc.

5,6,7,8-Tetrahydro-4-methylpyrido[3,4-d]pyrimidine hcl

  • MDL number MFCD11518992
  • Cas Number : 1187830-72-3
  • Molecular Weight : 185.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1603

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1603
Availability details for
Quantity
  • Availability
  • Add to Cart