COM448671468 Combi-Blocks, Inc.

1,8-Naphthyridine-2,7-diol

  • MDL number MFCD09258913
  • Cas Number : 49655-93-8
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1647

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1647
Availability details for
Quantity
  • Availability
  • Add to Cart