COM448671537 Combi-Blocks, Inc.

1-[(tert-Butyl)oxycarbonyl]-3-pyridin-2-ylmethylpiperidine-3-carboxylic acid

  • MDL number MFCD03840044
  • Cas Number : 887344-17-4
  • Molecular Weight : 320.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1724

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1724
Availability details for
Quantity
  • Availability
  • Add to Cart