COM448671548 Combi-Blocks, Inc.

1-[2-Amino-1-(2-methoxy-phenyl)-ethyl]-pyrrolidine-3-carboxylic acid

  • MDL number MFCD07369821
  • Cas Number : 886363-84-4
  • Molecular Weight : 264.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1735

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1735
Availability details for
Quantity
  • Availability
  • Add to Cart