COM448671568 Combi-Blocks, Inc.

13-Bromo-1-tridecanol

  • MDL number MFCD00482035
  • Cas Number : 116754-58-6
  • Molecular Weight : 279.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1762

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?SS-1762
Availability details for
Quantity
  • Availability
  • Add to Cart